Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Prodynorphin (Dynorphin B29), Mouse, Unlabeled, Clone: Dyn9,

Catalog No. MABT882MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT882MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT882MI Supplier MilliporeSigma™ Supplier No. MABT882
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Proenkephalin-B (UniProt P06300; also known as Beta-neoendorphin-dynorphin, Preprodynorphin) is encoded by the Pdyn gene (Gene ID 29190) in rat species. Proenkephalin B is an opioid polypeptide hormone involved with chemical signal transduction and cell communication. Prodynorphin is a basic building-block of endorphins, the chemical messengers in the brain that appear most heavily involved in the anticipation and experience of pain and the formation of deep emotional bonds critical in learning and memory. Prodynorphin is produced with a signal peptide (a.a. 1-21) and a propeptide (a.a. 22-163) sequence. Following the removal of the signal and propeptide sequences, the protein is further processed by proeolytic cleavages to form Alpha-neoendorphin (a.a. 166-175), Beta-neoendorphin (a.a. 166-174), Big dynorphin (a.a. 202-233; Big Dyn), Dynorphin A(1-17) (a.a. 202-218; Dyn-A17; Dynorphin A); Dynorphin A(1-13) (a.a. 202-214), Dynorphin A(1-8) (a.a. 202-209), Leu-enkephalin (a.a. 202-206 and 221-225), Leumorphin (Dynorphin B-29) (a.a. 221-248), and Rimorphin (a.a. 221-233; Dynorphin B; Dyn-B; Dynorphin B(1-13)).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prodynorphin (Dynorphin B29)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Dyn9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Pdyn
Gene Accession No. P06300
Host Species Mouse
Immunogen Thyroglobulin-conjugatged linear peptide corresponding to rat leumorphin (dynorphin B-29) C-terminal half sequence.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_062247
Test Specificity Clone Dyn9 recognizes preproenkephalin-B (a.a. 1-248 of rat sequence), proenkephalin-B (a.a. 22-248 of rat sequence), and leumorphin/dynorphin B-29 (a.a. 221-248 of rat sequence), but not other peptides derived from Proenkephalin-B.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.