Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prodynorphin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111373 Supplier Invitrogen™ Supplier No. PA5111373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLCAVKTQDG PKPINPLICS LQCQAALLPS EEWERCQSFL SFFTPSTLGL NDKEDLGSKS VGEGPYSELA KLSGSFLKEL EKSKFLPSIS TKE Highest antigen sequence indentity to the following orthologs: Mouse - 56%, Rat - 56%.

expression in hippocampus increases in response to traumatic brain injury; may play a role in regulatio of neurotransmitter release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prodynorphin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pdyn
Gene Accession No. P01213
Gene Alias ADCA; Alpha-neoendorphin; beta-neoendorphin; Beta-neoendorphin-dynorphin; Big Dyn; Big dynorphin; Dyn; Dyn-A17; Dyn-B; Dynorphin A(1-13); dynorphin A(1-17); Dynorphin A(1-8); Dynorphin B; Dynorphin B(1-13); Dynorphin B-29; Leu-enkephalin; leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; PDYN; PENKB; Preprodynorphin; preproenkephalin B; prodynorphin; proenkephalin B; proenkephalin-B; Rimorphin; SCA23; unnamed protein product
Gene Symbols Pdyn
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Prodynorphin. Recombinant protein control fragment (Product #RP-99329).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5173
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.