Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prodynorphin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA127973
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA127973 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA127973 Supplier Invitrogen™ Supplier No. PA127973
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

PA1-27973 detects ProDynorphin from guinea pig samples.

expression in hippocampus increases in response to traumatic brain injury; may play a role in regulatio of neurotransmitter release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prodynorphin
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene Pdyn
Gene Accession No. Q60478
Gene Alias ADCA; Alpha-neoendorphin; beta-neoendorphin; Beta-neoendorphin-dynorphin; Big Dyn; Big dynorphin; Dyn; Dyn-A17; Dyn-B; Dynorphin A(1-13); dynorphin A(1-17); Dynorphin A(1-8); Dynorphin B; Dynorphin B(1-13); Dynorphin B-29; Leu-enkephalin; leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; PDYN; PENKB; Preprodynorphin; preproenkephalin B; prodynorphin; proenkephalin B; proenkephalin-B; Rimorphin; SCA23; unnamed protein product
Gene Symbols Pdyn
Host Species Guinea Pig
Immunogen QEDPNAYSEEFFDV.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100135596
Target Species Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.