Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Professional HEALTHY SOAP™ Mild Foam
SDP

Catalog No. p-7145694 Shop All Gojo Industries, Inc. Products
Click to view available options
For Use With (Equipment):
PURELL ES6 Touch-Free Soap Dispensers
This item is not returnable. View return policy
This item is not returnable. View return policy

PURELL™ Professional HEALTHY SOAP™ Mild Foam for PURELL™ Soap Dispensers.

  • Exceptionally mild
  • Fragrance-free
  • Green-certified foam hand soap
  • Washes away dirt and germs
  • Enriched with a moisturizer and skin conditioners
  • Contains natural moisturizers
  • Nourishes the skin while cleaning
  • Supports your skin's natural ability to protect itself against dirt and germs
  • Hypoallergenic
  • AT-A-GLANCE™ refills make it obvious when refills need to be changed
  • SANITARY SEALED™ PET refill is easily recycled
TRUSTED_SUSTAINABILITY

Specifications

Product Type Foam Hand Wash
Color Blue
Certifications/Compliance Meets UL EcoLogo Standard for Hand Cleaner UL 2784, Meets USDA biobased Standard for hand cleaners
For Use With (Application) Hand Washing
For Use With (Equipment) PURELL ES6 Touch-Free Soap Dispensers
Quantity 2 Pk.
Volume (English) 40.58 oz.
Volume (Metric) 1200 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.