Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Profilin 1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299325MFV500
Description
Profilin 1 Polyclonal antibody specifically detects Profilin 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Profilin 1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
profilin 1, Profilin I, profilin-1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Profilin 1. | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
5216 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction