Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Profilin 2 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300301AF750
Description
Profilin 2 Polyclonal specifically detects Profilin 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Profilin 2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
D3S1319E, PFL, profilin 2, Profilin II, profilin-2 | |
Produced in rabbits immunized with purified, recombinant Profilin 2 (AAH18049.1; Met1-Phe140). | |
0.1 mL | |
Cell Biology, Cellular Markers, Signal Transduction, Stem Cell Markers | |
5217 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction