Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Profilin 2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NBP18742625UL

 View more versions of this product

Catalog No. NB406072


Only null left
Add to Cart

Description

Description

Profilin 2 Polyclonal specifically detects Profilin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Profilin 2
Polyclonal
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence validated from a verified customer review., Immunohistochemistry-Paraffin 1:50 - 1:200
D3S1319E, PFL, profilin 2, Profilin II, profilin-2
Rabbit
Affinity Purified
RUO
Primary
Specificity of human Profilin 2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
PFN2
This antibody was developed against Recombinant Protein corresponding to amino acids:QEAAIVGYCDAKYVWAATAGGVFQSITPIEIDMIVGKDREGFFTNGLTLGAKKCSVIRD
25 μL
Cytoskeleton Markers, Signal Transduction, Stem Cell Markers
5217
Human, Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only