Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Profilin 3 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5121475
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121475 100 μL
PIPA5121476 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121475 Supplier Invitrogen™ Supplier No. PA5121475
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PFN3 (Profilin 3) is a Protein Coding gene. Among its related pathways are Association Between Physico-Chemical Features and Toxicity Associated Pathways. Gene Ontology (GO) annotations related to this gene include actin binding and lipid binding. An important paralog of this gene is PFN1. The product of this gene belongs to the profilin family of proteins. This protein binds to actin and affects the structure of the cytoskeleton. It also may be involved in spermatogenesis. It is a single exon gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Profilin 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PFN3
Gene Accession No. P60673
Gene Alias Profilin III
Gene Symbols PFN3
Host Species Rabbit
Immunogen Met1-Arg133 (Accession P60673), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 345456
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.