Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ ProFlex™ 821 Smooth Surface Handling Work Gloves
Shop All Ergodyne Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Breathable polyester mesh keeps hands cool without being bulky or stiff
- Low-profile, molded hook & loop closure provides a secure fit
- Gloves feature an ID tab for personalization to prevent misplaced gloves
- Pull-on tab for easy on/off
- Machine washable
Specifications
Specifications
| Coating Material | Silicone |
| Color | Black |
| Hand | Specific |
| Material | Polyester Mesh |
| Product Type | Work Gloves |
| Size | Small |
| For Use With (Application) | Warehousing/Distribution, Package Delivery |
| Glove Cuff Style | Open |
| Series | ProFlex™ 821 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction