Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ ProFlex™ 4020 Wrist Support
Lightweight wrist support with open center is ideal for repetitive strain and overuse discomfort in office, retail and light industrial applications.
$138.00
Specifications
Color | Black |
---|---|
Size | 2X-Large |
For Use With (Application) | Food Processing, Picking/Packing,Assembly/Fabrication, Office/Retail, Light Industrial |
Catalog Number | Mfr. No. | Hand | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Hand | Price | Quantity & Availability | |||||
19-910-446
|
Ergodyne™
70208 |
Right |
Each for $138.00
|
|
|||||
19-910-447
|
Ergodyne™
70248 |
Left |
Each for $138.00
|
|
|||||
Specifications
Black | |
2X-Large | |
Food Processing, Picking/Packing,Assembly/Fabrication, Office/Retail, Light Industrial |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title