Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progerin Monoclonal Antibody (13A4), Active Motif™
SDP

Catalog No. 501993007
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3007 100 μg
50-199-3008 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3007 Supplier Active Motif Supplier No. 39965
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Progerin is a 614 amino acid protein involved in Hutchinson-Gilford progeria syndrome. Progerin is most often generated by a point mutation (C1824T) in the LMNA gene that codes for lamin-A/C. This mutation activates a cryptic splice site and gives rise to a form of lamin A with a 50-amino acid internal deletion within the C-terminal domain of the protein. Approximately 80% of Hutchinson-Gilford progeria syndrome cases carry a single copy of the most common mutation, a silent point mutation, G608G (GGC > GGT), within exon 11 of the LMNA gene.

Specifications

Antigen Progerin
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 13A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene LMNA
Gene Accession No. P02545
Gene Alias 70 kDa lamin; CDCD1; CDDC; CMD1A; CMT2B1; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; lamin; lamin A/C; lamin A/C-like 1; Lamin AC; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; LMNA; LMNC; LMNL1; Mutant lamin A/C; Prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32
Gene Symbols LMNA
Host Species Mouse
Immunogen This Progerin antibody was raised against a peptide corresponding to amino acids 604-611 of human Progerin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4000
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.