Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Progesterone Antibody (6-5E-10B), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253103JF646
Description
Progesterone Monoclonal antibody specifically detects Progesterone in All samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Progesterone | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
All species | |
Purified |
ELISA, Immunohistochemistry | |
6-5E-10B | |
ELISA, Immunohistochemistry | |
4-Pregnen-3,20-dione | |
Progesterone- 11a-hemisuccinate conjugated to bovine serum albumin (Pr11a-HMS-BSA) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction