Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Progesterone Monoclonal Antibody (2D9)
GREENER_CHOICE

Catalog No. PIMA549665 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549665 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549665 Supplier Invitrogen™ Supplier No. MA549665
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Progesterone is the major progestational steroid, secreted primarily by the corpus luteum and the placenta. Progesterone binds to the progesterone receptor, resulting in dissociation of heat shock proteins, receptor phosphorylation, and transcription activation through direct or indirect interaction with transcription factors. Progesterone acts on the uterus, the mammary glands and the brain. It is required in embryo implantation, pregnancy maintenance, and the development of mammary tissue for milk production. Progesterone, converted from pregnenolone, also serves as an intermediate in the biosynthesis of gonadal steroid hormones and adrenal corticosteroids. Therapeutic progesterone is a synthetic form of the endogenous hormone progesterone. This agent exerts inhibitory effects on estrogens by decreasing the number of estrogen receptors and increasing its metabolism to inactive metabolites. [National Center for Biotechnology Information. PubChem Compound Database; CID=5994].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Progesterone
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 2D9
Concentration 7.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias 11-alpha-hydroxyprogesterone; Agolutin; C21H30O2; Crinone; Luteohormone; OHP; Pregn-4-ene-3; Progesterone-11a-hemisuccinate
Host Species Mouse
Immunogen Conjugate of P hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.