Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Progesterone R/NR3C3 Antibody (OTI11E8), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273607R
Description
Progesterone R/NR3C3 Monoclonal specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Progesterone R/NR3C3 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PGR | |
Recombinant protein fragment corresponding to amino acids 1-298 of human Progesterone R/NR3C3 (NP_000917) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI11E8 | |
Western Blot, Immunohistochemistry | |
NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
Mouse | |
Affinity Purified | |
Breast Cancer, Cancer, Phospho Specific | |
5241.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction