Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Progesterone R/NR3C3 Antibody (PR501), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233369F
Description
Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Progesterone R/NR3C3 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PR501 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
Recombinant human Progesterone R/NR3C3 protein | |
0.1 mL | |
Breast Cancer, Cancer, Phospho Specific | |
5241 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction