Learn More
Epredia™ Lab Vision™ Progesterone Receptor Ab-14, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: PgR636
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: N-terminal (shared by A- and B-form)
Immunogen: Formalin fixed recombinant full-length A-form of human progesterone receptor
Molecular Weight: 116kDa (triplet, B-form) and 81kDa (singlet, A-form)
Positive Control: Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Progesterone Receptor Ab-14 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | PgR636 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Formalin fixed recombinant full length A-form of human progesterone receptor |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.