Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone Receptor Ab-8 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS298P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
1 mL
Form:
Concentrated
Concentration:
1mg/mL
200μg/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS298P1ABX 100 μL Concentrated 1mg/mL
MS298PABX 200 μL Concentrated 1mg/mL
MS298P 1 mL Concentrated 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS298P1ABX Supplier Epredia Supplier No. MS298P1ABX
Add to Cart
Add to Cart

Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-8, Mouse Monoclonal Antibody.

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 2 + hPRa 3

Isotype: IgG1/κ + IgG1/κ

Species Reactivity: Human, Horse, Sheep and Pig. Others not known.

Epitope: N-terminal half of human PgR

Immunogen: PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (B-form) and 81kDa (A-form)

Positive Control: T47D or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient

Specifications

Antigen Progesterone Receptor Ab-8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone hPRa 2+hPRa 3
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
Quantity 100 μL
Regulatory Status ASR, IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Equine, Human, Pig, Sheep
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.