Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progesterone Receptor (PR) Polyclonal Antibody, Abbomax

Catalog No. 502203643
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3643 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3643 Supplier Abbomax Inc Supplier No. 5002074
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Breast Cancer Tissue Cellular location: Nucleus, Cytoplasm.

Progesterone Receptor (PR) Polyclonal antibody specifically detects Progesterone Receptor (PR) in Bovine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot

Specifications

Antigen Progesterone Receptor (PR)
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene PGR
Gene Accession No. P06401, Q00175, Q63449
Gene Alias 9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human Progesterone receptor
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100848466, 18667, 25154, 5241
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.