Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Progestin Receptor Beta/MPRB Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981344
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1344 Supplier Bioss Supplier No. BS11410R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Progestin Receptor Beta-extra recognizes the extracellular epitope of the membrane progestin receptor. Progestin receptors with are found in pituitary, reproductive tract and most estrogen receptor-containing brain regions; these receptors are inducible by estrogen treatment. There are also progestin receptor sites in brain areas lacking estrogen receptors, such as the cerebral cortex of the rat; these receptors are not induced by estradiol treatment. Nevertheless, such receptors resemble those induced by estradiol. Another inducer of progestin receptors in brain is testosterone, which works through its conversion to estradiol via aromatization.

Specifications

Antigen Progestin Receptor Beta/MPRB
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PAQR8
Gene Accession No. Q8TEZ7
Gene Alias 1700019B16Rik; 3110001D06Rik; C6orf33; LMPB1; Lysosomal membrane protein in brain 1; lysosomal membrane protein in brain-1; Membrane progesterone P4 receptor beta; Membrane progesterone receptor beta; Membrane progestin receptor beta; mPR beta; MPRB; Paqr8; Pmrbeta; PR; Progesterone and adipoQ receptor family member 8; Progestin and adipoQ receptor family member 8; Progestin and adipoQ receptor family member VIII; progestin membrane receptor beta; putative membrane steroid receptor; RGD1311710
Gene Symbols PAQR8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Progestin Receptor Beta .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85315
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.