Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Proglumide sodium salt
GREENER_CHOICESDP

Catalog No. 147850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-785-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-785-0 Supplier Tocris Bioscience™ Supplier No. 1478/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-selective CCK antagonist; orally active

Non-selective cholecystokinin (CCK) antagonist. Inhibits CCK-stimulated amylase secretion and prevents CCK-induced 2-deoxyglucose uptake in mouse pancreatic acini. Blocks growth of HT29 colon carcinoma cells in response to gastrin 17 treatment. Orally active.

Chemical Identifiers

CAS 99247-33-3
Molecular Formula C18H25N2NaO4

Specifications

Quantity 50 mg
Formula Weight 356.39
Purity >99%
Molecular Weight (g/mol) 356.398
Chemical Name or Material Proglumide sodium salt
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.