Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Progranulin/PGRN Antibody (007), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289675IR
Description
Progranulin/PGRN Monoclonal antibody specifically detects Progranulin/PGRN in Human samples. It is validated for ELISASpecifications
Progranulin/PGRN | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
007 | |
ELISA | |
CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Progranulin/PGRN (NP_002078.1; Met 1-Leu 593). | |
0.1 mL | |
Cancer, Lipid and Metabolism | |
2896 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction