Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Progranulin/PGRN Antibody (034), Alexa Fluor™ 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290563AF350
Description
Progranulin/PGRN Monoclonal specifically detects Progranulin/PGRN in Mouse samples. It is validated for ELISA.Specifications
Progranulin/PGRN | |
Monoclonal | |
Alexa Fluor 350 | |
CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Progranulin/PGRN (NP_032201.2; Met 1-Leu 589). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human GRN | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
34 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
2896 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction