Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Progranulin/PGRN Rabbit anti-Mouse, Clone: 34, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00
Specifications
Antigen | Progranulin/PGRN |
---|---|
Clone | 34 |
Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Progranulin/PGRN Monoclonal specifically detects Progranulin/PGRN in Mouse samples. It is validated for ELISA, Sandwich ELISA Capture.Specifications
Progranulin/PGRN | |
ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
2896 | |
IgG | |
Protein A purified |
34 | |
ELISA | |
Unconjugated | |
Rabbit | |
Mouse | |
CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN | |
Recombinant Mouse Progranulin/PGRN protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title