Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prohibitin 2 Antibody [DyLight 550], Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals AF4794L
Description
Prohibitin 2 Polyclonal antibody specifically detects Prohibitin 2 in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
Prohibitin 2 | |
Polyclonal | |
50mM Sodium Borate | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
DyLight 550 | |
BAP, Bap37, BCAP37, B-cell associated protein, D-prohibitin, MGC117268, p22, PNAS-141, prohibitin 2, prohibitin-2, REAB-cell receptor-associated protein BAP37, Repressor of estrogen receptor activity | |
E. coli-derived recombinant human Prohibitin 2, Lys89-Lys299, Accession # Q99623 | |
0.1 mL | |
Cellular Markers, Signal Transduction | |
11331 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction