Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prohibitin 2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001807H
Description
Prohibitin 2 Polyclonal antibody specifically detects Prohibitin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Prohibitin 2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BAP, Bap37, BCAP37, B-cell associated protein, D-prohibitin, MGC117268, p22, PNAS-141, prohibitin 2, prohibitin-2, REAB-cell receptor-associated protein BAP37, Repressor of estrogen receptor activity | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Repressor of Estrogen Activity (Prohibitin 2) using the numbering given in entry NP_009204.1 (GeneID 11331). | |
0.1 mL | |
Cellular Markers, Signal Transduction | |
11331 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction