Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prohibitin 2 Antibody (SR1516), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32214750UL
Description
Prohibitin 2 Monoclonal antibody specifically detects Prohibitin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
Prohibitin 2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1516 | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
BAP, Bap37, BCAP37, B-cell associated protein, D-prohibitin, MGC117268, p22, PNAS-141, prohibitin 2, prohibitin-2, REAB-cell receptor-associated protein BAP37, Repressor of estrogen receptor activity | |
A synthesized peptide derived from human Prohibitin 2 (Uniprot #: Q99623) | |
50 μg | |
Cellular Markers, Signal Transduction | |
11331 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction