Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prohibitin 2 Mouse anti-Human, Mouse, Rat, Clone: 1D9C7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Prohibitin 2 |
---|---|
Clone | 1D9C7 |
Concentration | 0.7 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6851
![]() ![]() |
Proteintech Group Inc
664241IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PHB2 acts as a mediator of transcriptional repression by nuclear hormone receptors via recruitment of histone deacetylases. This protein functions as an estrogen receptor (ER)-selective coregulator that potentiates the inhibitory activities of antiestrogens and represses the activity of estrogens. It competes with NCOA1 for modulation of ER transcriptional activity. It is probably involved in regulating mitochondrial respiration activity and in aging.Specifications
Prohibitin 2 | |
0.7 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BAP, Bap37, D prohibitin, p22, PHB2, PNAS 141, prohibitin 2, REA | |
PHB2 | |
IgG1 | |
Protein G | |
Antibody |
1D9C7 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
O35129, Q5XIH7, Q99623 | |
11331, 114766, 12034 | |
Prohibitin 2 Fusion Protein Ag2977 | |
Primary | |
-20°C | |
PHB2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title