Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prohibitin Antibody (SPM311), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308722C
Description
Prohibitin Monoclonal antibody specifically detects Prohibitin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)Specifications
Prohibitin | |
Monoclonal | |
DyLight 650 | |
PHB1, prohibitin | |
Recombinant human Prohibitin protein fragment (aa167-261) (exact sequence is proprietary) (Uniprot: P35232) | |
100 μg | |
Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
5245 | |
Store at 4°C in the dark. | |
IgG2c κ |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
SPM311 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction