Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prohibitin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587348 Supplier Invitrogen™ Supplier No. PA587348
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Raji, NIH/3T3, HeLa, MCF7, HepG2, Mouse liver, Mouse heart, Mouse kidney, Rat heart; Cellular Location: Mitochondrion inner membrane Immunogen sequence: PRNVPVITGS KDLQNVNITL RILFRPVASQ LPRIFTSIGE DYDERVLPSI TTEILKSVVA RFDAGELITQ RELVSRQVSD DLTERAATFG LILDDVSLTH LTFGKEFTEA VEAKQVAQQE AERARFVVEK AEQQKKAAII SAEGDSKAAE LIANSLATAG DGLIELRKLE AAEDIAYQLS RSRNITYLPA GQSVLLQLPQ.

Prohibitin is from an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3' untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3' untranslated region may function as a trans-acting regulatory RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prohibitin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PHB
Gene Accession No. P35232, P67778, P67779
Gene Alias BAP 32; Bap32; B-cell receptor-associated protein 32; epididymis luminal protein 215; epididymis secretory sperm binding protein Li 54e; fa18f07; fb09f05; HEL-215; HEL-S-54e; OTTHUMP00000165765; OTTHUMP00000165766; OTTHUMP00000220103; OTTHUMP00000220170; OTTHUMP00000220171; PHB; phb protein; PHB1; Prohibitin; sr:nyz128; wu:fa18f07; wu:fb09f05
Gene Symbols PHB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 73-272 of human Prohibitin (NP_0026251).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18673, 25344, 5245
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.