Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prohibitin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595085
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595085 Supplier Invitrogen™ Supplier No. PA595085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Lung Tissue, Rat Skeletal Muscle Tissue, Rat Brain Tissue, Rat Kidney Tissue, HELA Cell, MCF-7 Cell, PC-12 Cell, A549 Cell, SMMC Cell, mouse NIH3T3 whole cell. IHC: Human Lung Cancer Tissue, rat brain tissue. ICC/IF: U20S cell, HELA Cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Prohibitin is from an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3' untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3' untranslated region may function as a trans-acting regulatory RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prohibitin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene PHB
Gene Accession No. P35232, P67778, P67779
Gene Alias BAP 32; Bap32; B-cell receptor-associated protein 32; epididymis luminal protein 215; epididymis secretory sperm binding protein Li 54e; fa18f07; fb09f05; HEL-215; HEL-S-54e; OTTHUMP00000165765; OTTHUMP00000165766; OTTHUMP00000220103; OTTHUMP00000220170; OTTHUMP00000220171; PHB; phb protein; PHB1; Prohibitin; sr:nyz128; wu:fa18f07; wu:fb09f05
Gene Symbols PHB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human Prohibitin (240-256aa KLEAAEDIAYQLSRSRN).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18673, 25344, 5245
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.