Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Prohibitin Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $482.00
Specifications
Antigen | Prohibitin |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119357
|
Invitrogen™
PA5119357 |
200 μL |
Each for $482.00
|
|
|||||
PIPA5121668
|
Invitrogen™
PA5121668 |
20 μL |
Each for $178.40
|
|
|||||
Description
Prohibitin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Prohibitin is from an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3' untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3' untranslated region may function as a trans-acting regulatory RNA.Specifications
Prohibitin | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Mouse, Rat | |
PHB | |
BAP 32; Bap32; B-cell receptor-associated protein 32; epididymis luminal protein 215; epididymis secretory sperm binding protein Li 54e; fa18f07; fb09f05; HEL-215; HEL-S-54e; OTTHUMP00000165765; OTTHUMP00000165766; OTTHUMP00000220103; OTTHUMP00000220170; OTTHUMP00000220171; PHB; phb protein; PHB1; Prohibitin; sr:nyz128; wu:fa18f07; wu:fb09f05 | |
PHB | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P35232, P67778, P67779 | |
18673, 25344, 5245 | |
Recombinant protein Prohibitin. The antigen corresponds to amino acid range 177-272 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title