Learn More
Prohibitin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00585T

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Rabbit, Guinea pig_10141, Pig, Panda, Cat, Orangutan, Monkey, Gor
Prohibitin is an evolutionarily conserved gene that is ubiquitously expressed. It is thought to be a negative regulator of cell proliferation and may be a tumor suppressor. Mutations in PHB have been linked to sporadic breast cancer. Prohibitin is expressed as two transcripts with varying lengths of 3′ untranslated region. The longer transcript is present at higher levels in proliferating tissues and cells, suggesting that this longer 3′ untranslated region may function as a trans-acting regulatory RNA.Specifications
Prohibitin | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P35232 | |
PHB | |
Between 222 and 272 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
PHB | |
PHB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5245 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.