Learn More
                                    Proinsulin Antibody (253627), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB13361MFV500
Description
Proinsulin Monoclonal antibody specifically detects Proinsulin in Human, Mouse samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| Proinsulin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology | |
| 3630 | |
| Store at 4C in the dark. | 
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 253627 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| E. coli-derived recombinant human Proinsulin, Phe25-Asn110, Accession # P01308 | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2a | 