Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proinsulin Antibody (C-PEP-01), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500413B
Description
Proinsulin Monoclonal antibody specifically detects Proinsulin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune AssaysSpecifications
Proinsulin | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
C-PEP-01 | |
ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Radioimmunoassay | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
C-peptide (Human) conjugated to BSA. | |
0.1 mL | |
Diabetes Research, Immune System Diseases, Immunology | |
3630 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction