Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proinsulin Antibody (C-PEP-01) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500413
Description
Proinsulin Monoclonal specifically detects Proinsulin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Radioimmunoassay.Specifications
Proinsulin | |
Monoclonal | |
1.0 mg/mL | |
ELISA 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 2 to 5 μg/mL, Radioimmunoassay 1:100-1:2000 | |
INS | |
C-peptide (Human) conjugated to BSA. | |
0.1 mg | |
Diabetes Research, Immune System Diseases, Immunology | |
3630 | |
Human | |
IgG1 |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
C-PEP-01 | |
Unconjugated | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The antibody C-PEP-01 reacts specifically with C-peptide, a part of the Proinsulin molecule. Proinsulin consists of the three parts: C-peptide and two long strands of amino acids (alpha and beta chains; later become linked together to form the Insulin molecule). No cross-reactivity with Insulin or other peptide hormones or proteins was observed. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction