Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PROK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567568 Supplier Invitrogen™ Supplier No. PA567568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PROK1 Polyclonal Antibody for IHC (P)

Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROK1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PROK1
Gene Accession No. P58294
Gene Alias black mamba toxin-related protein; EGVEGF; EG-VEGF; endocrine-gland-derived vascular endothelial growth factor; Mambakine; PK1; PRK1; PROK1; prokineticin 1; prokineticin-1; UNQ600/PRO1186
Gene Symbols PROK1
Host Species Rabbit
Immunogen Synthesized peptide derived from human PROK1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84432
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.