Learn More
PROK1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
$2175.00
Specifications
Antigen | PROK1 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-271-8625
![]() |
Invitrogen™
500P188GBT500UG |
500 μg |
Each of 1 for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: AVITGACERD VQCGAGTCCA ISLWLRGLRM CTPLGREGEE CHPGSHKVPF FRKRKHHTCP CLPNLLCSRF PDGRYRCSMD LKNINF. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human EG-VEGF. Anti-Human EG-VEGF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hEG-VEGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human EG-VEGF (500-P188G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hEG-VEGF. Western Blot: To detect Human EG-VEGF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human EG-VEGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P188GBT-1MG will be provided as 2 x 500 μg
Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.Specifications
PROK1 | |
ELISA, Western Blot | |
Biotin | |
Goat | |
Human | |
P58294 | |
84432 | |
E.coli-derived Recombinant Human EG-VEGF. | |
Antigen affinity chromatography | |
PROK1 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
black mamba toxin-related protein; EGVEGF; EG-VEGF; endocrine-gland-derived vascular endothelial growth factor; Mambakine; PK1; PRK1; PROK1; prokineticin 1; prokineticin-1; UNQ600/PRO1186 | |
PROK1 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.