Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROK2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986800
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6800 Supplier Bioss Supplier No. BS5784R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Prokineticin-2 (PK2) is a cysteine-rich secreted protein that is expressed in the suprachiasmatic nucleus (SCN) with receptors located in the critical autonomic control centers of the brain. It has a depolarizing effect on neurons expressing the receptor. PK2 is predominantly controlled by the endogenous circadian clock, but light also plays a modulatory role. PK2 functions as a critical SCN output molecule responsible for circadian locomotor rhythms. PK2 expression is high during the day, and responsive to nocturnal light pulses. PK2 also functions as a chemoattractant for subventricular zone-derived neuronal progenitors.

Specifications

Antigen PROK2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PROK2
Gene Accession No. Q9HC23
Gene Alias Bombina variegata 8 kD protein; Bombina variegata 8kDa protein; BOS_20837; Bv 8 homolog (Bombina variegata); Bv8; HH4; homolog of mouse Bv8 (Bombina variegata 8 kDa); homolog of mouse Bv8 (Bombina variegata 8 kDa); prokineticin 2; KAL4; MIT1; PK2; Prok1; PROK2; prokineticin 1; prokineticin 2; prokineticin 2 beta; Prokineticin-2; Protein Bv8 homolog
Gene Symbols PROK2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PROK2/Prokineticin 2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60675
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.