Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prokineticin R1/PROKR1 Antibody (420849), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4655APCCY7
Description
Prokineticin R1/PROKR1 Monoclonal antibody specifically detects Prokineticin R1/PROKR1 in Human samples. It is validated for Flow CytometrySpecifications
Prokineticin R1/PROKR1 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
420849 | |
Flow Cytometry | |
G protein-coupled receptor 73, G protein-coupled receptor ZAQ, GPR73, GPR73aG-protein coupled receptor 73, PK-R1, PKR1G-protein coupled receptor ZAQ, prokineticin receptor 1, ZAQ | |
NS0 mouse myeloma cell line transfected with human Prokineticin R1/PKR1, Met1-Lys393, Accession # Q8TCW9 | |
0.1 mL | |
GPCR | |
10887 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction