Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prokineticin R2/PROKR2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP145218
Description
Prokineticin R2/PROKR2 Polyclonal specifically detects Prokineticin R2/PROKR2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Prokineticin R2/PROKR2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
dJ680N4.3, G protein-coupled receptor 73-like 1, GPR73bG-protein coupled receptor 73-like 1, GPR73L1Kallmann syndrome 3 (autosomal dominant), GPRg2KAL3, PKR2PK-R2, prokineticin receptor 2 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody recognizes and epitope within the internal region. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3-1 ug/ml, Immunohistochemistry 3.75 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_658986.1 | |
PROKR2 | |
Peptide with sequence C-NGNTSFTPNFN corresponding to internal region according to NP_658986.1. | |
0.1 mg | |
Protein Phosphatase | |
128674 | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction