Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prokineticin Receptor 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519208
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519208 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519208 Supplier Invitrogen™ Supplier No. PA519208
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Prokineticins are secreted proteins that can promote angiogenesis and induce strong gastrointestinal smooth muscle contraction. GPR73B is an integral membrane protein and G protein-coupled receptor for prokineticins. The encoded protein is similar in sequence to GPR73, another G protein-coupled receptor for prokineticins. GPCR73B expression has been reported in brain, ovary, salivary gland, small intestine (ileocecum), testis, and thyroid. No human ESTs have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prokineticin Receptor 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PROKR2
Gene Accession No. Q8NFJ6, Q8R415
Gene Alias B830005M06Rik; dJ680N4.3; EG-VEGRF2; G protein-coupled receptor 73-like 1; G protein-coupled receptor I5E; GPCR73B; Gpcr73l1; GPR73b; GPR73L1; GPRg2; G-protein coupled receptor 73-like 1; G-protein coupled receptor I5E; HH3; KAL3; OTTHUMP00000030192; PKR2; PK-R2; Prokineticin receptor 2; Prokr2
Gene Symbols PROKR2
Host Species Goat
Immunogen Synthetic peptide sequence (NGNTSFTPNFN) corresponding to the internal amino acids of PROKR2 (aa 5-15).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128674, 192649
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.