Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prokineticin Receptor 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534110
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534110 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534110 Supplier Invitrogen™ Supplier No. PA534110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon (100%) Monkey, Marmoset (93%) Horse (87%) Elephant, Rabbit (80%).

Prokineticins are secreted proteins that can promote angiogenesis and induce strong gastrointestinal smooth muscle contraction. GPR73B is an integral membrane protein and G protein-coupled receptor for prokineticins. The encoded protein is similar in sequence to GPR73, another G protein-coupled receptor for prokineticins. GPCR73B expression has been reported in brain, ovary, salivary gland, small intestine (ileocecum), testis, and thyroid. No human ESTs have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prokineticin Receptor 2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PROKR2
Gene Accession No. Q8NFJ6
Gene Alias B830005M06Rik; dJ680N4.3; EG-VEGRF2; G protein-coupled receptor 73-like 1; G protein-coupled receptor I5E; GPCR73B; Gpcr73l1; GPR73b; GPR73L1; GPRg2; G-protein coupled receptor 73-like 1; G-protein coupled receptor I5E; HH3; KAL3; OTTHUMP00000030192; PKR2; PK-R2; Prokineticin receptor 2; Prokr2
Gene Symbols PROKR2
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from N-terminal extracellular domain of human PROKR2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128674
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.