Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prokineticin Receptor 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534111
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534111 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534111 Supplier Thermo Scientific Supplier No. PA534111
Add to Cart
Add to Cart

Prokineticin Receptor 2 Polyclonal Antibody

Prokineticin Receptor 2 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Prokineticin Receptor 2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene PROKR2
Gene Accession No. Q8NFJ6
Gene Alias B830005M06Rik; dJ680N4.3; EG-VEGRF2; G protein-coupled receptor 73-like 1; G protein-coupled receptor I5E; GPCR73B; Gpcr73l1; GPR73b; GPR73L1; GPRg2; G-protein coupled receptor 73-like 1; G-protein coupled receptor I5E; HH3; KAL3; OTTHUMP00000030192; PKR2; PK-R2; Prokineticin receptor 2; Prokr2
Gene Symbols PROKR2
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human PROKR2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128674
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.