Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prolactin Antibody (CL6553), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Prolactin |
---|---|
Clone | CL6553 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Prolactin Monoclonal specifically detects Prolactin in Human samples. It is validated for Western Blot.Specifications
Prolactin | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
prolactin | |
PRL | |
IgG2a | |
Protein A purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL6553 | |
Monoclonal | |
Purified | |
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
5617.0 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: TEVRGMQEAPEAILSKAVEIEEQTKRLLEGMELIVSQVHPETKENEIYPVWSGLPSLQMADEESRLSAYYNLLHCLRRDSHKIDNYLKLLKCRIIHNNN | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title