Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prolactin Antibody (INN-hPRL-1), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062210AF350
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
Prolactin | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Ion exchange chromatography | |
RUO | |
Primary | |
Human | |
IgG1 |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
INN-hPRL-1 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
prolactin | |
Made to Human PROLACTIN | |
0.25 mL | |
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction | |
5617 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction