Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prolactin Antibody (OTI6B1), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273609APCCY7
Description
Prolactin Monoclonal specifically detects Prolactin in Human samples. It is validated for Flow Cytometry.Specifications
Prolactin | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
PRL | |
Full length recombinant protein of human Prolactin (NP_000939) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Flow Cytometry | |
OTI6B1 | |
Flow Cytometry | |
prolactin | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction | |
5617.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction