Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Prolactin Antibody (OTI6B1), Allophycocyanin, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273609APC
Description
Prolactin Monoclonal specifically detects Prolactin in Human samples. It is validated for Flow Cytometry.Specifications
| Prolactin | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| PRL | |
| Full length recombinant protein of human Prolactin (NP_000939) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a | 
| Flow Cytometry | |
| OTI6B1 | |
| Flow Cytometry | |
| prolactin | |
| Mouse | |
| Affinity Purified | |
| Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction | |
| 5617.0 | |
| Store at 4°C in the dark. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction