Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prolactin Antibody (OTI6B1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273609H
Description
Prolactin Monoclonal specifically detects Prolactin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Prolactin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PRL | |
Full length recombinant protein of human Prolactin (NP_000939) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI6B1 | |
Western Blot, Immunohistochemistry | |
prolactin | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction | |
5617.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction