Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prolactin Antibody (PRL/4908R), DyLight 755, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308249IR
Description
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Prolactin | |
Monoclonal | |
DyLight 755 | |
prolactin | |
Recombinant fragment of human Prolactin (PRL) protein (around aa 63-201) (exact sequence is proprietary) (Uniprot: IDP01236) | |
100 μg | |
Biologically Active Proteins, Cancer, Signal Transduction | |
5617 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PRL/4908R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction