Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prolactin Antibody (rPRL/4909) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308245
Description
Prolactin Monoclonal specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Prolactin | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
prolactin | |
Recombinant fragment of human Prolactin protein (around aa 63-201) (exact sequence is proprietary) | |
100 μg | |
Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction | |
5617 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rPRL/4909 | |
Unconjugated | |
10mM PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction